Learn More
3-Bromo-vortioxetine Hydrobromide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-vortioxetine Hydrobromide |
| CAS | 2725536-51-4 |
| Molecular Formula | C18H21BrN2S • HBr |
| Quantity | 100 mg |
| Synonym | 1-(2-((3-Bromo-2,4-dimethylphenyl)thio)phenyl)piperazine Hydrobromid |
| InChI Formula | InChI=1S/C18H21BrN2S.BrH/c1-13-7-8-16(14(2)18(13)19)22-17-6-4-3-5-15(17)21-11-9-20-10-12-21;/h3-8,20H,9-12H2,1-2H3;1H |
| SMILES | CC1=C(SC2=CC=CC=C2N3CCNCC3)C=CC(C)=C1Br.Br |
| IUPAC Name | 1-[2-(3-bromo-2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide |
| Molecular Weight (g/mol) | 458.25 |
| Formula Weight | 455.99 |