Learn More
3-Bromo Nevirapine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo Nevirapine |
| CAS | 284686-21-1 |
| Molecular Formula | C15H13BrN4O |
| Quantity | 100 mg |
| Synonym | 3-Bromo-11-cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| InChI Formula | InChI=1S/C15H13BrN4O/c1-8-11(16)7-18-14-12(8)19-15(21)10-3-2-6-17-13(10)20(14)9-4-5-9/h2-3,6-7,9H,4-5H2,1H3,(H,19,21) |
| SMILES | Cc1c(Br)cnc2N(C3CC3)c4ncccc4C(=Nc12)O |
| IUPAC Name | 6-bromo-2-cyclopropyl-7-methyl-2,4,9,15-tetrazatricyclo[9.4.0.03,8]pentadeca-1(11),3,5,7,12,14-hexaen-10-one |
| Molecular Weight (g/mol) | 345.19 |
| Formula Weight | 344.0273 |