Learn More
3-Bromo-5-(1-hydroxyethyl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-5-(1-hydroxyethyl)pyridine |
| CAS | 283608-66-2 |
| Molecular Formula | C7H8BrNO |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C7H8BrNO/c1-5(10)6-2-7(8)4-9-3-6/h2-5,10H,1H3 |
| SMILES | CC(O)c1cncc(Br)c1 |
| IUPAC Name | 1-(5-bromopyridin-3-yl)ethanol |
| Molecular Weight (g/mol) | 202.05 |
| Formula Weight | 200.9789 |