Learn More
3-(Boc-aminomethyl)azetidine hydrochloride, 95%
Product Code. 15425286
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Product Code. 15425286
Brand: Thermo Scientific Alfa Aesar H59816.ME
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 91188-15-7 | C9H18N2O2 | 186.26 g/mol
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 91188-15-7
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD01861760
InChI Key MOLUHRBHXXGWDP-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine
PubChem CID 15346872
SMILES CC(C)(C)OC(=O)NCC1CNC1
Specifications
Solubility Information Slightly soluble in water.
IUPAC Name tert-butyl N-[(azetidin-3-yl)methyl]carbamate
Quantity 500 mg
Formula Weight 222.71
Percent Purity 95%
Chemical Name or Material 3-(Boc-aminomethyl)azetidine hydrochloride