missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, 99%, Thermo Scientific™

Product Code. p-7047825
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329785 250 mg 250mg
11319785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329785 Supplier Thermo Scientific Alfa Aesar Supplier No. L19807.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 284493-53-4
Molecular Formula C15H20NO5
Molecular Weight (g/mol) 294.33
MDL Number MFCD02090693
InChI Key DNXONWHJJNOKEU-LBPRGKRZSA-M
Synonym 3-boc-amino-3-3-methoxyphenyl propionic acid, 3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, 3-n-boc-3-3-methoxyphenyl propionic acid, 3-tert-butoxy carbonylamino-3-3-methoxyphenyl propanoic acid, acmc-1cl2c, 3-3-methoxyphenyl-3-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-3-amino-3-3-methoxyphenyl propionic acid, 3-boc-amino-3-3-methoxyphenyl propanoic acid, 3-tert-butoxy carbonyl amino-3-3-methoxyphenyl propanoic acid
PubChem CID 2734591
IUPAC Name 3-(3-methoxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES COC1=CC=CC(=C1)[C@H](CC([O-])=O)NC(=O)OC(C)(C)C

Specifications

Melting Point 81°C to 83°C
Quantity 250 mg
Formula Weight 295.34
Percent Purity 99%
Chemical Name or Material 3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.