Learn More
3-Benzyloxy-4-methoxy-Beta-nitrostyrene, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Benzyloxy-4-methoxy-beta-nitrostyrene |
| CAS | 55507-05-6 |
| Molecular Formula | C16 H15 N O4 |
| Quantity | 1 g |
| Synonym | 3-Benzyloxy-4-Methoxy-Ss-Nitrostyrene |
| InChI Formula | InChI=1S/C16H15NO4/c1-20-15-8-7-13(9-10-17(18)19)11-16(15)21-12-14-5-3-2-4-6-14/h2-11H,12H2,1H3/b10-9+ |
| SMILES | COc1ccc(\C=C\[N+](=O)[O-])cc1OCc2ccccc2 |
| IUPAC Name | 1-methoxy-4-[(E)-2-nitroethenyl]-2-phenylmethoxybenzene |
| Molecular Weight (g/mol) | 285.29 |
| Formula Weight | 285.1001 |