Learn More
3-(Benzyl(butyl)amino)propanoic Acid, TRC
Product Code. 30347565
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347565 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347565 Supplier Toronto Research Chemicals Supplier No. TRCB21825150MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(Benzyl(butyl)amino)propanoic Acid
CAS 216587-41-6
Molecular Formula C14H21NO2
Quantity 50 mg
InChI Formula InChI=1S/C14H21NO2/c1-2-3-10-15(11-9-14(16)17)12-13-7-5-4-6-8-13/h4-8H,2-3,9-12H2,1H3,(H,16,17)
SMILES CCCCN(CCC(=O)O)Cc1ccccc1
IUPAC Name 3-[benzyl(butyl)amino]propanoic acid
Molecular Weight (g/mol) 235.32
Formula Weight 235.1572