Learn More
(3-Aminopropyl)trimethylazanium Hydrochloride Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | (3-Aminopropyl)trimethylazanium Hydrochloride Chloride |
| CAS | 3399-68-6 |
| Molecular Formula | C6H17N2. HCl . Cl |
| Quantity | 10 mg |
| Synonym | 3-Amino-N,N,N-trimethyl-1-propanaminium Chloride Hydrochloride |
| InChI Formula | InChI=1S/C6H17N2.2ClH/c1-8(2,3)6-4-5-7;;/h4-7H2,1-3H3;2*1H/q+1;;/p-1 |
| SMILES | Cl.[Cl-].C[N+](C)(C)CCCN |
| IUPAC Name | 3-aminopropyl(trimethyl)azanium;chloride;hydrochloride |
| Molecular Weight (g/mol) | 189.12 |
| Formula Weight | 188.0847 |