Learn More
(3-Aminopropyl)ethyl-carbamic Acid tert-Butyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (3-Aminopropyl)ethyl-carbamic Acid tert-Butyl Ester |
| CAS | 273409-54-4 |
| Molecular Formula | C10H22N2O2 |
| Quantity | 100 mg |
| Synonym | (3-Aminopropyl)ethyl-carbamic Acid 1,1-Dimethylethyl Ester, tert-Butyl N-(3-Aminopropyl)-N-(ethyl)carbamate |
| InChI Formula | InChI=1S/C10H22N2O2/c1-5-12(8-6-7-11)9(13)14-10(2,3)4/h5-8,11H2,1-4H3 |
| SMILES | CCN(CCCN)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-(3-aminopropyl)-N-ethylcarbamate |
| Molecular Weight (g/mol) | 202.29 |