Learn More
3-Amino-3-deoxy-D-mannose Hydrochloride, TRC
Product Code. 30179202
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179202 500 mg 500mg
30179373 100 mg 100mg
30179057 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179202 Supplier Toronto Research Chemicals Supplier No. TRCA604000500MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69880-85-9
Molecular Formula C6 H13 N O5 . Cl H
Molecular Weight (g/mol) 215.63
IUPAC Name (2S,3S,4S,5R)-3-amino-2,4,5,6-tetrahydroxyhexanal;hydrochloride
SMILES Cl.N[C@H]([C@H](O)C=O)[C@H](O)[C@H](O)CO
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C6H13NO5.ClH/c7-5(3(10)1-8)6(12)4(11)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4-,5-,6-;/m1./s1
Formula Weight 215.0561
Chemical Name or Material 3-Amino-3-deoxy-D-mannose, Hydrochloride