Learn More
3-Amino-2-methyl-2-propanol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-2-methyl-2-propanol |
| CAS | 2854-16-2 |
| Molecular Formula | C4H11NO |
| Quantity | 10 g |
| Synonym | ,1-Dimethylethanolamine, 1-Amino-2-methyl-2-propanol, 2-Amino-alpha,alpha-dimethylethanol, 2-Hydroxy-2-methyl-1-propylamine, 2-Hydroxy-2-methylpropan-1-amine, 2-Hydroxyisobutylamine, 2-Methyl-2-hydroxypropylamine, NSC 17697 |
| InChI Formula | InChI=1S/C4H11NO/c1-4(2,6)3-5/h6H,3,5H2,1-2H3 |
| SMILES | CC(C)(O)CN |
| IUPAC Name | 1-amino-2-methylpropan-2-ol |
| Molecular Weight (g/mol) | 89.14 |
| Formula Weight | 89.0841 |