Learn More
3-Amino-2,2-dimethylpropanamide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-2,2-dimethylpropanamide |
| CAS | 324763-51-1 |
| Molecular Formula | C5H12N2O |
| Quantity | 100 mg |
| Synonym | 3-Amino-2,2-dimethyl-propanamide, 3-Amino-2,2-dimethylpropionamide |
| InChI Formula | InChI=1S/C5H12N2O/c1-5(2,3-6)4(7)8/h3,6H2,1-2H3,(H2,7,8) |
| SMILES | CC(C)(CN)C(=O)N |
| IUPAC Name | 3-amino-2,2-dimethylpropanamide |
| Molecular Weight (g/mol) | 116.16 |
| Formula Weight | 116.095 |