Learn More
3-Amino-1,2,4-benzotriazine 1,4-Dioxide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-1,2,4-benzotriazine-1,4-dioxide |
| CAS | 27314-97-2 |
| Molecular Formula | C7 H6 N4 O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C7H6N4O2/c8-7-9-11(13)6-4-2-1-3-5(6)10(7)12/h1-4H,(H2,8,9) |
| SMILES | Nc1n[n+]([O-])c2ccccc2[n+]1[O-] |
| IUPAC Name | 1,4-dioxido-1,2,4-benzotriazine-1,4-diium-3-amine |
| Molecular Weight (g/mol) | 178.15 |
| Formula Weight | 178.0491 |