Learn More
3,6-Dichloroacenaphthene, TRC
Product Code. 30374715
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374715 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30374715 Supplier Toronto Research Chemicals Supplier No. TRCD4315752.5MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,6-Dichloroacenaphthene
CAS 19202-59-6
Molecular Formula C12H8Cl2
Quantity 2.5 mg
Synonym 3,6-Dichloro-1,2-dihydroacenaphthylene
InChI Formula InChI=1S/C12H8Cl2/c13-10-5-2-7-1-3-8-11(14)6-4-9(10)12(7)8/h2,4-6H,1,3H2
SMILES Clc1ccc2c(Cl)ccc3CCc1c23
IUPAC Name 3,6-dichloro-1,2-dihydroacenaphthylene
Molecular Weight (g/mol) 223.1
Formula Weight 222.0003