missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-4-methylpyridazine, 97%, Thermo Scientific Chemicals

Product Code. p-7024784
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11430441

Brand: Thermo Scientific Alfa Aesar A19998.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19064-64-3
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006465
InChI Key ROYHWGZNGMXQEU-UHFFFAOYSA-N
Synonym 4-methyl-3,6-dichloropyridazine, 3,6-dichloro-4-methyl-pyridazine, pyridazine, 3,6-dichloro-4-methyl, 3,6-dichloro-5-methylpyridazine, 3,6-dichloro-4-methyl-1,2-diazine, pubchem9497, ksc495e6l, 3,6-dichloro-4-methylpyridazin, pyridazine,6-dichloro-4-methyl, 3,6-di chloro-4-methyl pyridazine
PubChem CID 87923
IUPAC Name 3,6-dichloro-4-methylpyridazine
SMILES CC1=CC(=NN=C1Cl)Cl

Specifications

Melting Point 86°C to 88°C
Boiling Point 149°C to 151°C (21 mmHg)
Quantity 1 g
Beilstein 119573
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 3,6-Dichloro-4-methylpyridazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.