Learn More
3,6-Dichloro-2-methoxy-d3-benzoic Acid, CDN
Product Code. 30321595
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321595 0.005 g 0.01g
30319679 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321595 Supplier CDN Supplier No. CDND40850.005G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dicamba-d3
CAS 349553-95-3
Molecular Formula C8 2H3 H3 Cl2 O3
Quantity 0.005 g
Synonym Benzoic acid, 3,6-dichloro-2-(methoxy-d3)- (9 CI), Dicamba D3 (methoxy D3), Dicamba-d3
InChI Formula InChI=1 S/C8H6Cl2O3/c1-13-7-5(10)3-2-4(9)6(7)8(11)12/h2-3 H,1H3,(H,11,12)/i1D3
SMILES [2 H]C([2 H])([2 H])Oc1c(Cl)ccc(Cl)c1C(=O)O
IUPAC Name 3,6-dichloro-2-(trideuteriomethoxy)benzoic acid
Molecular Weight (g/mol) 224.06
Formula Weight 222.9882 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity