Learn More
3-(5-Methyl-2-furyl)butyraldehyde, 99%
Product Code. 11405307
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405307 5 g 5g
11415307 25 g 25g
11425307 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11405307 Supplier Thermo Scientific Alfa Aesar Supplier No. B22671.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 31704-80-0 | C9H12O2 | 152.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31704-80-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00209519
InChI Key PFLUFBMKOXFWSD-UHFFFAOYNA-N
Synonym 3-5-methyl-2-furyl butyraldehyde, 3-5-methylfuran-2-yl butanal, 3-5-methyl-2-furyl butanal, 3-5-methyl-2-furanyl butanal, fema no. 3307, beta5-dimethyl-2-furanpropanal, 2-furanpropanal, beta5-dimethyl, 2-furanpropanal, beta,5-dimethyl, beta5-dimethyl-2-furanpropionaldehyde, 2-furanpropanal, .beta.,5-dimethyl
PubChem CID 3578033
IUPAC Name 3-(5-methylfuran-2-yl)butanal
SMILES CCC(C=O)C1=CC=C(C)O1
Tekniske data
Density 1.006
Boiling Point 93°C to 94°C (14 mmHg)
Flash Point 58°C (136°F)
Quantity 5 g
UN Number UN1989
Sensitivity Air Sensitive
Formula Weight 152.19
Percent Purity 99%
Chemical Name or Material 3-(5-Methyl-2-furyl)butyraldehyde
RUO – Research Use Only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.