Learn More
3,5-Diiodo Thyroaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Diiodo Thyroaldehyde |
| CAS | 2828-49-1 |
| Molecular Formula | C13H8I2O3 |
| Quantity | 100 mg |
| Synonym | 4-(4-Hydroxyphenoxy)-3,5-diiodo-benzaldehyde |
| InChI Formula | InChI=1S/C13H8I2O3/c14-11-5-8(7-16)6-12(15)13(11)18-10-3-1-9(17)2-4-10/h1-7,17H |
| SMILES | Oc1ccc(Oc2c(I)cc(C=O)cc2I)cc1 |
| IUPAC Name | 4-(4-hydroxyphenoxy)-3,5-diiodobenzaldehyde |
| Molecular Weight (g/mol) | 466.01 |
| Formula Weight | 465.8563 |