Learn More
3,5-Difluorocinnamic acid, 98%, Thermo Scientific™
Product Code. 11419467
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419467 1 g 1g
11429467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11419467 Supplier Thermo Scientific Alfa Aesar Supplier No. B21991.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84315-23-1
Molecular Formula C9H6F2O2
Molecular Weight (g/mol) 184.142
MDL Number MFCD00010321
InChI Key MBAWRXICVNIUGY-OWOJBTEDSA-N
Synonym 3,5-difluorocinnamic acid, trans-3,5-difluorocinnamic acid, 3,5-difluorocinnamicacid, e-3-3,5-difluorophenyl acrylic acid, 3-3,5-difluorophenyl acrylic acid, 2e-3-3,5-difluorophenyl prop-2-enoic acid, 2e-3-3,5-difluorophenyl acrylic acid, rarechem bk hw 0255, timtec-bb sbb006675, 3-3,5-difluorophenyl propenoic acid
PubChem CID 5374941
IUPAC Name (E)-3-(3,5-difluorophenyl)prop-2-enoic acid
SMILES C1=C(C=C(C=C1F)F)C=CC(=O)O
Specifications
Melting Point 204°C to 205°C
Quantity 1 g
Beilstein 6194926
Formula Weight 184.14
Percent Purity 98%
Chemical Name or Material 3,5-Difluorocinnamic acid