Learn More
3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%
Product Code. 15448587
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15448587
Brand: Thermo Scientific Alfa Aesar H52069.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 205526-24-5 | C24H19F2NO4 | 423.42 g/mol
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 205526-24-5
Molecular Formula C24H19F2NO4
Molecular Weight (g/mol) 423.42
MDL Number MFCD00797579
InChI Key UYEQBZISDRNPFC-QFIPXVFZSA-N
Synonym fmoc-phe 3,5-f2-oh, fmoc-phe 3,5-dif-oh, fmoc-3,5-difluoro-l-phenylalanine, s-n-fmoc-3,5-difluorophenylalanine, fmoc-l-3,5-difluorophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3,5-difluorophenyl propanoic acid, fmoc-l-3,5-difluorophe, 3,5-difluoro-l-phenylalanine, n-fmoc protected, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3,5-difluorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2774201
SMILES OC(=O)[C@H](CC1=CC(F)=CC(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Specifications
Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-3-(3,5-difluorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Formula Weight 423.4
Percent Purity 95%
Chemical Name or Material 3,5-Difluoro-N-Fmoc-L-phenylalanine