Learn More
3,5-Difluoro-L-phenylalanine, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Difluoro-L-phenylalanine |
| CAS | 31105-91-6 |
| Molecular Formula | C9 H9 F2 N O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H9F2NO2/c10-6-1-5(2-7(11)4-6)3-8(12)9(13)14/h1-2,4,8H,3,12H2,(H,13,14)/t8-/m0/s1 |
| SMILES | N[C@@H](Cc1cc(F)cc(F)c1)C(=O)O |
| IUPAC Name | (2S)-2-amino-3-(3,5-difluorophenyl)propanoic acid |
| Molecular Weight (g/mol) | 201.17 |
| Formula Weight | 201.06 |