Learn More
3,5-Difluoro-4-methoxybenzyl bromide, 97%
Product Code. 11485930
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11485930 5 g 5g
11475930 1 g 1g
2 options
Product Code. 11485930
Brand: Thermo Scientific Alfa Aesar H26526.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 706786-42-7 | C8H7BrF2O | 237.04 g/mol
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 706786-42-7
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.04
MDL Number MFCD04115916
InChI Key IQTBMXJVMUSGSL-UHFFFAOYSA-N
Synonym 3,5-difluoro-4-methoxybenzyl bromide, 5-bromomethyl-1,3-difluoro-2-methoxybenzene, 4-bromomethyl-2,6-difluoroanisole, benzene, 5-bromomethyl-1,3-difluoro-2-methoxy
PubChem CID 17750664
IUPAC Name 5-(bromomethyl)-1,3-difluoro-2-methoxybenzene
SMILES COC1=C(F)C=C(CBr)C=C1F
Specifications
Melting Point 39°C to 42°C
Quantity 5 g
UN Number UN3261
Solubility Information Insoluble in water.
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-4-methoxybenzyl bromide