Learn More
3,5-Dichloroiodobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Dichloroiodobenzene |
| CAS | 3032-81-3 |
| Molecular Formula | C6H3Cl2I |
| Quantity | 500 mg |
| Synonym | 3,5-Dichlorophenyl Iodide, 3,5-Dichloro-1-iodobenzene, 1,3-Dichloro-5-iodobenzene, 1-Iodo-3,5-dichlorobenzene |
| InChI Formula | InChI=1S/C6H3Cl2I/c7-4-1-5(8)3-6(9)2-4/h1-3H |
| SMILES | Clc1cc(Cl)cc(I)c1 |
| IUPAC Name | 1,3-dichloro-5-iodobenzene |
| Molecular Weight (g/mol) | 272.89 |
| Formula Weight | 271.8656 |