Learn More
3,5-Dichloroacenaphthene, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Dichloroacenaphthene |
| CAS | 27608-78-2 |
| Molecular Formula | C12H8Cl2 |
| Quantity | 25 mg |
| Synonym | 3,5-Dichloro-1,2-dihydroacenaphthylene |
| InChI Formula | InChI=1S/C12H8Cl2/c13-10-6-11(14)9-5-4-7-2-1-3-8(10)12(7)9/h1-3,6H,4-5H2 |
| SMILES | Clc1cc(Cl)c2cccc3CCc1c23 |
| IUPAC Name | 3,5-dichloro-1,2-dihydroacenaphthylene |
| Molecular Weight (g/mol) | 223.1 |
| Formula Weight | 222.0003 |