Learn More
3,5-Dibromo-4-hydroxybenzaldehyde, 97%
Product Code. 11435123
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435123 5 g 5g
11445123 25 g 25g
11455123 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11435123 Supplier Thermo Scientific Alfa Aesar Supplier No. A12780.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 2973-77-5 | C7H4Br2O2 | 279.92 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2973-77-5
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00016980
InChI Key SXRHGLQCOLNZPT-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dibromo-4-hydroxy, timtec-bb sbb006529, acmc-1cr6c, 3,5-dibromo-4-hydroxy benzaldehyde, 3,5-dibromo-4-hydroxy-benzaldehyde, 4-hydroxy-3,5-dibromo benzaldehyde, 3,5-dibromo-4-hydroxybenzaldehyde, 3,5-bis bromanyl-4-oxidanyl-benzaldehyde
PubChem CID 18100
IUPAC Name 3,5-dibromo-4-hydroxybenzaldehyde
SMILES OC1=C(Br)C=C(C=O)C=C1Br
Specifications
Melting Point 179°C to 182°C
Quantity 5 g
Beilstein 1948512
Sensitivity Air Sensitive
Formula Weight 279.93
Percent Purity 98%
Chemical Name or Material 3,5-Dibromo-4-hydroxybenzaldehyde