missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Bromo-2-methoxy-3-pyridyl)acrylic acid, Thermo Scientific™

Product Code. p-7033105
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494560 1 g 1g
11404570 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494560 Supplier Thermo Scientific Alfa Aesar Supplier No. H50117.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1228963-18-5
Molecular Formula C9H8BrNO3
Molecular Weight (g/mol) 258.071
MDL Number MFCD07781178
InChI Key AJOCJTKIVWCVMY-NSCUHMNNSA-N
Synonym 3-5-bromo-2-methoxypyridin-3-yl acrylic acid, 3-5-bromo-2-methoxy-3-pyridyl acrylic acid, 3-5-bromo-2-methoxy-pyridin-3-yl-acrylic acid, 2e-3-5-bromo-2-methoxypyridin-3-yl acrylic acid, e-3-5-bromo-2-methoxypyridin-3-yl acrylic acid, 2e-3-5-bromo-2-methoxy 3-pyridyl prop-2-enoic acid, 2e-3-5-bromo-2-methoxypyridin-3-yl prop-2-enoic acid
PubChem CID 42553143
IUPAC Name (E)-3-(5-bromo-2-methoxypyridin-3-yl)prop-2-enoic acid
SMILES COC1=NC=C(C=C1C=CC(=O)O)Br

Spezifikation

Quantity 1 g
Formula Weight 258.07
Chemical Name or Material 3-(5-Bromo-2-methoxy-3-pyridyl)acrylic acid
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.