missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propylamine, 98%

Product Code. p-7027040
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11435487

Brand: Thermo Scientific Alfa Aesar B23485.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-00-2 | C7H16N2O | 144.22 g/mol

3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-00-2
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD00006184
InChI Key UIKUBYKUYUSRSM-UHFFFAOYSA-N
Synonym n-3-aminopropyl morpholine, 4-morpholinepropanamine, 3-morpholinopropylamine, 3-morpholinopropan-1-amine, n-aminopropylmorpholine, 4-3-aminopropyl morpholine, 4-morpholinepropylamine, 4-aminopropylmorpholine, 1-amino-3-morpholinopropane, 3-4-morpholinyl-1-propanamine
PubChem CID 61055
ChEBI CHEBI:86554
IUPAC Name 3-morpholin-4-ylpropan-1-amine
SMILES NCCCN1CCOCC1

Specifications

Melting Point -15°C
Density 0.987
Boiling Point 223°C to 225°C
Flash Point 98°C (210°F)
Refractive Index 1.4761
Quantity 50 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinyl)propylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-(4-Morpholinyl)propylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : N-Aminopropylmorpholine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.