Learn More
3,4-Methylenedioxy Propiophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Methylenedioxy Propiophenone |
| CAS | 28281-49-4 |
| Molecular Formula | C10 H10 O3 |
| Quantity | 5 g |
| Synonym | 1-Propanone','1-(1,3-benzodioxol-5-yl)-, Propiophenone','3',4'-(methylenedioxy)- (6CI,7CI,8CI), 1-(1,3-Benzodioxol-5-yl)-1-propanone, 1-(2H-1,3-Benzodioxol-5-yl)propan-1-one, 1-(Benz[d][1,3]dioxol-5-yl)propan-1-one, 1-(Benzo[d][1,3]dioxol-5-yl)propan-1-one, 3,4-Methylenedioxyphenyl ethyl ketone, 3,4-Methylenedioxypropiophenone, 3',4'-(Methylenedioxy)propiophenone, 5-Propanoyl-1,3-benzodioxole, NSC 29484, 3,4-Methylenedioxy propiophenone |
| InChI Formula | InChI=1S/C10H10O3/c1-2-8(11)7-3-4-9-10(5-7)13-6-12-9/h3-5H,2,6H2,1H3 |
| SMILES | CCC(=O)c1ccc2OCOc2c1 |
| IUPAC Name | 1-(1,3-benzodioxol-5-yl)propan-1-one |
| Molecular Weight (g/mol) | 178.18 |
| Formula Weight | 178.063 |