Learn More
3,4-Dimethoxybenzenesulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Dimethoxybenzenesulfonyl Chloride |
| CAS | 23095-31-0 |
| Molecular Formula | C19 H10 N2 O |
| Quantity | 10 g |
| Synonym | 3,4-Bis(methoxy)benzenesulfonyl Chloride, 3,4-Dimethoxybenzene-1-sulfonyl Chloride, 3,4-Dimethoxyphenylsulfonyl Chloride |
| InChI Formula | InChI=1S/C19H10N2O/c22-10-14-18-12-6-2-4-8-16(12)20-17(18)9-13-11-5-1-3-7-15(11)21-19(13)14/h1-10H |
| SMILES | O=CC1=C2C(=Nc3ccccc23)C=C4C1=Nc5ccccc45 |
| IUPAC Name | indolo[3,2-b]carbazole-12-carbaldehyde |
| Molecular Weight (g/mol) | 282.296 |
| Formula Weight | 282.079 |