Learn More
3,4-Dimethoxybenzamidoxime, 97%, Thermo Scientific™
Product Code. 15458777
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458777 5 g 5g
15448777 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15458777 Supplier Thermo Scientific Alfa Aesar Supplier No. H52203.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1053058-99-3
Molecular Formula C9H12N2O3
Molecular Weight (g/mol) 196.21
MDL Number MFCD06147747,MFCD09842202
InChI Key REKFOHFJHBBXOR-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzamidoxime, z-n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide, 3,4-dimethoxyphenyl hydroxyimino methylamine, n'-hydroxy-3,4-dimethoxybenzimidamide, benzenecarboximidamide, n-hydroxy-3,4-dimethoxy, n'-hydroxy-3,4-dimethoxybenzene-1-carboximidamide, n'∼1∼-hydroxy-3,4-dimethoxy-1-benzenecarboximidamide, e-n'-hydroxy-3,4-dimethoxybenzenecarboximidamide
PubChem CID 9723450
IUPAC Name N'-hydroxy-3,4-dimethoxybenzenecarboximidamide
SMILES COC1=C(OC)C=C(C=C1)C(N)=NO
Specifications
Quantity 5 g
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzamidoxime