Learn More
(3,4-Dihydroxyphenyl)-2-pyridinyl-methanone Hydrobromide, TRC
Specifications
Specifications
| Chemical Name or Material | (3,4-Dihydroxyphenyl)-2-pyridinyl-methanone Hydrobromide |
| CAS | 31749-12-9 |
| Molecular Formula | C12H9NO3.HBr |
| Quantity | 10 mg |
| Synonym | 3,4-Dihydroxyphenyl 2-Pyridyl Ketone Hydrobromide |
| InChI Formula | InChI=1S/C12H9NO3.BrH/c14-10-5-4-8(7-11(10)15)12(16)9-3-1-2-6-13-9;/h1-7,14-15H;1H |
| SMILES | Br.Oc1ccc(cc1O)C(=O)c2ccccn2 |
| IUPAC Name | (3,4-dihydroxyphenyl)-pyridin-2-ylmethanone;hydrobromide |
| Molecular Weight (g/mol) | 296.11 |
| Formula Weight | 294.9844 |