Learn More
3,4-Dihydroxybenzoic Acid Methyl Ester-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 3,4-Dihydroxybenzoic Acid Methyl Ester-d3 |
| CAS | 2734001-51-3 |
| Molecular Formula | C8 D3 H5 O4 |
| Quantity | 10 mg |
| Synonym | Protocatechuic Acid Methyl-d3 Ester, 3,4-Dihydroxy MetDyl-d3 Benzoate, Methyl-d3 3,4-dihydroxybenzoate, Methyl-d3 Protocatechuate, NSC 146458-d3 |
| InChI Formula | InChI=1S/C8H8O4/c1-12-8(11)5-2-3-6(9)7(10)4-5/h2-4,9-10H,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])OC(=O)c1ccc(O)c(O)c1 |
| IUPAC Name | trideuteriomethyl 3,4-dihydroxybenzoate |
| Molecular Weight (g/mol) | 171.165 |
| Formula Weight | 171.061 |