missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzophenone, 98%, Thermo Scientific™

Product Code. 11454374
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454374 5 g 5g
11464374 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454374 Supplier Thermo Scientific Alfa Aesar Supplier No. A13067.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85118-07-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00009892
InChI Key ZJTYHSBOZAQQGF-UHFFFAOYSA-N
Synonym 3,4-difluorobenzophenone, 3,4-difluorophenyl phenyl methanone, methanone, 3,4-difluorophenyl phenyl, 3,4-difluoro-phenyl-phenyl-methanone, 3,4-difluorophenyl phenyl ketone, fr bf dvr, pubchem4220, acmc-209q4q, timtec-bb sbb016583
PubChem CID 569908
IUPAC Name (3,4-difluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)F)F

Specifications

Melting Point 55°C to 57°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 7808356
Formula Weight 218.2
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.