Learn More
3,4-Dichloro-6-fluoroaniline, 96%
Product Code. 15475848
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475848 250 mg 250mg
15465848 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15475848 Supplier Thermo Scientific Alfa Aesar Supplier No. H59506.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 2729-36-4 | C6H4Cl2FN | 180.003 g/mol
3,4-Dichloro-6-fluoroaniline is used as pharmaceutical intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dichloro-6-fluoroaniline is used as pharmaceutical intermediates
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2729-36-4
Molecular Formula C6H4Cl2FN
Molecular Weight (g/mol) 180.003
MDL Number MFCD19707630
InChI Key FHJVHVKVTUTGTA-UHFFFAOYSA-N
Synonym 3,4-dichloro-6-fluoroaniline, 4,5-dichloro-2-fluorobenzenamine, 4,5-dichloro-2-fluorophenylamine, 4,5-dichloro-2-fluoro-phenylamine, 4,5-dichloro-2-fluorophenyl amine, benzenamine, 4,5-dichloro-2-fluoro
PubChem CID 223091
IUPAC Name 4,5-dichloro-2-fluoroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)F)N
Specifications
Quantity 250 mg
UN Number UN2811
Formula Weight 180.01
Percent Purity 96%
Chemical Name or Material 3,4-Dichloro-6-fluoroaniline