Learn More
3,4,5-Trifluorobenzonitrile, 97%, Thermo Scientific™
Product Code. 11486366
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486366 5 g 5g
11476366 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11486366 Supplier Thermo Scientific Alfa Aesar Supplier No. B23143.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 134227-45-5
Molecular Formula C7H2F3N
Molecular Weight (g/mol) 157.095
MDL Number MFCD00074948
InChI Key XFKYJMGXZXJYBS-UHFFFAOYSA-N
Synonym 3,4,5-trifluoro-1-cyano benzene, benzonitrile, 3,4,5-trifluoro, 3,4,5-trifluoro-benzonitrile, 3,4,5-trifluorobenzenecarbonitrile, pubchem1573, intermediates-zcf02038, acmc-1bz57, ksc493s4j, 3,4,5-?trifluorobenzonitrile, 3,4,5-trifluorobenzonitrile
PubChem CID 593814
IUPAC Name 3,4,5-trifluorobenzonitrile
SMILES C1=C(C=C(C(=C1F)F)F)C#N
Specifications
Melting Point 44°C to 47°C
Boiling Point 72°C to 73°C (10 mmHg)
Flash Point 62°C (143°F)
Quantity 5 g
UN Number UN3439
Beilstein 7702244
Formula Weight 157.1
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzonitrile