Learn More
3,4,5-Triethoxybenzoic acid, 98+%
Product Code. 11462197
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462197 25 g 25g
11472197 100 g 100g
11482197 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11462197 Supplier Thermo Scientific Alfa Aesar Supplier No. A14157.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 6970-19-0 | C13H18O5 | 254.282 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6970-19-0
Molecular Formula C13H18O5
Molecular Weight (g/mol) 254.282
MDL Number MFCD00002505
InChI Key YCENSLDYFDZUMO-UHFFFAOYSA-N
Synonym benzoic acid,3,4,5-triethoxy, benzoic acid, 3,4,5-triethoxy, 3,4,5-triethoxy benzoic acid, acmc-1b720
PubChem CID 81432
IUPAC Name 3,4,5-triethoxybenzoic acid
SMILES CCOC1=CC(=CC(=C1OCC)OCC)C(=O)O
Specifications
Melting Point 110°C to 113°C
Quantity 25 g
Beilstein 2659063
Formula Weight 254.29
Percent Purity ≥98%
Chemical Name or Material 3,4,5-Triethoxybenzoic acid