missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric acid, 98+%

Product Code. p-7025068
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11434307

Brand: Thermo Scientific Alfa Aesar B20388.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4839-46-7 | C7H12O4 | 160.17 g/mol

It acts as a reactant involved in cyclodehydration of diols, synthesis of conjugates of betulin derivatives used as anti-HIV agents, preparation of dimeric peptide antagonists of IgG-FcRn interaction, microwave-assisted protection of glutaraldehyde, synthesis of glycyrrhetinic acid derivatives for proteasome inhibition and catalytic, asymmetric transannular aldolizations. It is also used with other dibasic esters as solvents in paints, cosmetics, and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a reactant involved in cyclodehydration of diols, synthesis of conjugates of betulin derivatives used as anti-HIV agents, preparation of dimeric peptide antagonists of IgG-FcRn interaction, microwave-assisted protection of glutaraldehyde, synthesis of glycyrrhetinic acid derivatives for proteasome inhibition and catalytic, asymmetric transannular aldolizations. It is also used with other dibasic esters as solvents in paints, cosmetics, and fragrances.

Solubility
Solubility in water, very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4839-46-7
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00002716
InChI Key DUHQIGLHYXLKAE-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid
PubChem CID 20984
ChEBI CHEBI:68502
IUPAC Name 3,3-dimethylpentanedioic acid
SMILES CC(C)(CC(O)=O)CC(O)=O

Specifications

Melting Point 98°C to 103°C
Density 1.42
Boiling Point 89°C to 90°C (2 mmHg)
Quantity 10 g
Beilstein 1765439
Formula Weight 160.17
Percent Purity ≥98%
Chemical Name or Material 3,3-Dimethylglutaric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.