Learn More
3,3-Dimethyl-2,4-pentanedione, TRC
Product Code. 30381723
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381723 1 g 1g
30371871 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30381723 Supplier Toronto Research Chemicals Supplier No. TRCD4768451G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3142-58-3
Molecular Formula C20 H35 F15 O5 Si5
Molecular Weight (g/mol) 780.893
Synonym 3,3-Dimethylacetylacetone, NSC 42609
IUPAC Name 2,4,6,8,10-pentamethyl-2,4,6,8,10-pentakis(3,3,3-trifluoropropyl)-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
SMILES C[Si]1(CCC(F)(F)F)O[Si](C)(CCC(F)(F)F)O[Si](C)(CCC(F)(F)F)O[Si](C)(CCC(F)(F)F)O[Si](C)(CCC(F)(F)F)O1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C20H35F15O5Si5/c1-41(11-6-16(21,22)23)36-42(2,12-7-17(24,25)26)38-44(4,14-9-19(30,31)32)40-45(5,15-10-20(33,34)35)39-43(3,37-41)13-8-18(27,28)29/h6-15H2,1-5H3
Formula Weight 780.109
Chemical Name or Material 3,3-Dimethyl-2,4-pentanedione