Learn More
3,3'-Diiodo-L-thyronine, TRC
Product Code. 30010421
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010421 50 mg 50mg
30005029 100 mg 100mg
17945035 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010421 Supplier Toronto Research Chemicals Supplier No. TRCD45515050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4604-41-5
Molecular Formula C15 H13 I2 N O4
Molecular Weight (g/mol) 525.08
Synonym L-Tyrosine, O-(4-hydroxy-3-iodophenyl)-3-iodo-, Alanine, 3-[4-(4-hydroxy-3-iodophenoxy)-3-iodophenyl]-, L- (8CI), 3,3'-Diiodo-L-thyronine, L-3,3'-Diiodothyronine, O-(4-Hydroxy-3-iodophenyl)-3-iodo-L-tyrosine, Levothyroxine Sodium Imp J (EP)
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3-iodophenyl]propanoic acid
SMILES N[C@@H](Cc1ccc(Oc2ccc(O)c(I)c2)c(I)c1)C(=O)O
Specifications
InChI Formula InChI=1S/C15H13I2NO4/c16-10-7-9(2-3-13(10)19)22-14-4-1-8(5-11(14)17)6-12(18)15(20)21/h1-5,7,12,19H,6,18H2,(H,20,21)/t12-/m0/s1
Quantity 50 mg
Formula Weight 524.8934
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3,3'-Diiodo-L-thyronine
Grade HPLC