Learn More
MedChemExpress 3,3'-Dihexyloxacarbocyanine iodide
Product Code. 16434667
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16434667 Supplier MedChemExpress Supplier No. HYD0084
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
MedChemExpress 3,3'-Dihexyloxacarbocyanine iodide is a carbocyanine dye which can be used to monitor changes in mitochondrial membrane potential.
Specifications
Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : 33.33 mg/mL (58.22 mM; Need ultrasonic)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 572.52 g/mol
CAS 53213-82-4
Synonym DiOC6(3) iodide