missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromo-4-methoxyphenyl)propionic acid, 96%, Thermo Scientific™

Product Code. p-7031826
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418285 250 mg 250mg
15475827 1 g 1g
15485827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418285 Supplier Thermo Scientific Alfa Aesar Supplier No. H33042.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1857-57-4
Molecular Formula C10H11BrO3
Molecular Weight (g/mol) 259.10
MDL Number MFCD00184737
InChI Key MLXVZRVWVCOUBQ-UHFFFAOYSA-N
Synonym 3-3-bromo-4-methoxyphenyl propanoic acid, benzenepropanoicacid, 3-bromo-4-methoxy, 3-3-bromo-4-methoxyphenyl propionic acid, benzenepropanoic acid, 3-bromo-4-methoxy, 3-3-bromo-4-methoxy-phenyl-propionic acid
PubChem CID 4547608
IUPAC Name 3-(3-bromo-4-methoxyphenyl)propanoic acid
SMILES COC1=C(Br)C=C(CCC(O)=O)C=C1

Specifications

Melting Point 96°C to 100°C
Quantity 250 mg
Formula Weight 259.1
Percent Purity 96%
Chemical Name or Material 3-(3-Bromo-4-methoxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.