Learn More
3-(3-Aminopropyl)benzoic Acid Hydrochloride, TRC
Product Code. 30313872
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313872 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30313872 Supplier Toronto Research Chemicals Supplier No. TRCA62824550MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(3-Aminopropyl)benzoic Acid Hydrochloride
CAS 1346604-68-9
Molecular Formula C10H14ClNO2
Quantity 50 mg
Synonym Fesoterodine Impurity 3 HCl
InChI Formula InChI=1S/C10H13NO2.ClH/c11-6-2-4-8-3-1-5-9(7-8)10(12)13;/h1,3,5,7H,2,4,6,11H2,(H,12,13);1H
SMILES C1=CC(=CC(=C1)CCCN)C(=O)O.Cl
IUPAC Name 3-(3-aminopropyl)benzoic acid hydrochloride
Molecular Weight (g/mol) 215.68
Formula Weight 215.07