Learn More
3,3',5,5'-Tetrabromobisphenol A-d10, (OH)2, TRC
Product Code. 30416186
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416186 50 mg -
30427524 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30416186 Supplier Toronto Research Chemicals Supplier No. TRCT29110950MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2732834-96-5
Molecular Formula C15 D10 H2 Br4 O2
Molecular Weight (g/mol) 553.9322
Synonym Tetrabromobisphenol A D10 (dimethyl D3, bisphenol-3,5-D2), Tetrabromobisphenol A D10, 4,4'-(1,1,1,3,3,3-Hexadeuterio-2,2-propanediyl)bis(2,6-dibromo-3,5-dideuteriophenol), 2,2',6,6'-Tetrabromobisphenol A (ring,chain-D10), 3,3',5,5'-Tetrabromobisphenol A (ring,dimethyl-D10), 3,3',5,5'-Tetrabromobisphenol A D10 (ring D4 dimethyl D6)
IUPAC Name 2,6-dibromo-3,5-dideuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(3,5-dibromo-2,6-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol
SMILES [2H]c1c(Br)c(O)c(Br)c([2H])c1C(c2c([2H])c(Br)c(O)c(Br)c2[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3/i1D3,2D3,3D,4D,5D,6D
Formula Weight 549.8198
Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A-d10