Learn More
3,3',5,5'-Tetrabromobisphenol A-d10, (OH)2, CDN
Product Code. 30325932
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325932 0.01 g 0.01g
30323415 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325932 Supplier CDN Supplier No. CDND79330.01G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A-d10
CAS 2732834-96-5
Molecular Formula C15 D10 H2 Br4 O2
Quantity 0.01 g
Synonym Tetrabromobisphenol A D10 (dimethyl D3, bisphenol-3,5-D2);Tetrabromobisphenol A D10, 4,4'-(1,1,1,3,3,3-Hexadeuterio-2,2-propanediyl)bis(2,6-dibromo-3,5-dideuteriophenol), 2,2',6,6'-Tetrabromobisphenol A (ring,chain-D10), 3,3',5,5'-Tetrabromobisphenol A (ring,dimethyl-D10), 3,3',5,5'-Tetrabromobisphenol A D10 (ring D4 dimethyl D6)
InChI Formula InChI=1 S/C15H12Br4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D
SMILES [2 H]c1c(Br)c(O)c(Br)c([2 H])c1C(c2c([2 H])c(Br)c(O)c(Br)c2[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 2,6-dibromo-3,5-dideuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(3,5-dibromo-2,6-dideuterio-4-hydroxyphenyl)propan-2-yl]phenol
Molecular Weight (g/mol) 553.9322
Formula Weight 549.8198 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity