Learn More
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanesulfonic Acid, Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1,2,2H-Perfluorodecanesulfonic acid sodium salt |
| CAS | 27619-96-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H4 F17 O3 S . Na |
| Quantity | 25 mg |
| Synonym | 1H,1H,2H,2H-Perfluorodecanesulfonic acid sodium, 1-Decanesulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, sodium salt (1:1), 1-Decanesulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, sodium salt (8CI,9CI) |
| InChI Formula | InChI=1S/C10H5F17O3S.Na/c11-3(12,1-2-31(28,29)30)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27;/h1-2H2,(H,28,29,30);/q;+1/p-1 |
| SMILES | [Na+].[O-]S(=O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | sodium;3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecane-1-sulfonate |
| Molecular Weight (g/mol) | 550.16 |
| Show More |