Learn More
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane-1-sulphonic Acid, TRC
Product Code. 17914985
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914985 25 mg 25mg
17903735 100 mg 100mg
17924645 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17914985 Supplier Toronto Research Chemicals Supplier No. TRCT00780525MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27619-97-2
Molecular Formula C8 H5 F13 O3 S
Molecular Weight (g/mol) 428.17
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanesulfonic acid, 1,1,2,2-Tetrahydroperfluorooctane sulfonate, 1H,1H,2H,2H-Perfluorooctanesulfonic acid, 2-(Perfluorohexyl)ethane-1-sulfonic acid, 2-(Perfluorohexyl)ethanesulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane-1-sulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctanesulfonic acid, THPFOS, 6:2 FTSA
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctane-1-sulfonic acid
SMILES OS(=O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C8H5F13O3S/c9-3(10,1-2-25(22,23)24)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h1-2H2,(H,22,23,24)
Formula Weight 427.9752
Percent Purity >95%
Chemical Name or Material 1,1,2,2H-Perfluorooctanesulfonic acid
Grade HPLC