Learn More
3-(2-Methylphenyl)propionic acid, 96%
Product Code. 15467307
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467307 1 g 1g
15477307 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15467307 Supplier Thermo Scientific Alfa Aesar Supplier No. H34375.03
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 22084-89-5 | C10H12O2 | 164.204 g/mol
3-(2-Methylphenyl)propionic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(2-Methylphenyl)propionic acid is a carboxylic acid building block. It participates in the synthesis of 2-methyl-3-phenylpropanol.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22084-89-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00079773
InChI Key JIRKNEAMPYVPTD-UHFFFAOYSA-N
PubChem CID 30938
IUPAC Name 3-(2-methylphenyl)propanoic acid
SMILES CC1=CC=CC=C1CCC(=O)O
Specifications
Melting Point 101°C to 103°C
Quantity 1 g
Synonym 3-2-methylphenyl propionic acid, 3-2-methylphenyl propanoic acid, 2-methylhydrocinnamic acid, benzenepropanoic acid, 2-methyl, 3-o-tolyl propanoic acid, 3-o-tolylpropanoic acid, 2-methylbenzenepropanoic acid, benzenepropanoic acid,2-methyl, 2-methylhydrozimtsaure
Solubility Information Soluble in water.
Formula Weight 164.2
Percent Purity 96%
Chemical Name or Material 3-(2-Methylphenyl)propionic acid