Learn More
3’-(2-Fluorophenyl) Ezetimibe, TRC
Specifications
Specifications
| Chemical Name or Material | 3'-(2-Fluorophenyl) Ezetimibe |
| CAS | 2743547-96-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24H21F2NO3 |
| Quantity | 1 mg |
| Synonym | Ezetimibe Impurity, (3R,4S)-1-(4-Fluorophenyl)-3-[(3S)-3-(2-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone |
| InChI Formula | InChI=1S/C24H21F2NO3/c25-16-7-9-17(10-8-16)27-23(15-5-11-18(28)12-6-15)20(24(27)30)13-14-22(29)19-3-1-2-4-21(19)26/h1-12,20,22-23,28-29H,13-14H2/t20-,22+,23-/m0/s1 |
| SMILES | O[C@@H](CC[C@@H]1[C@H](N(C1=O)c2ccc(F)cc2)c3ccc(O)cc3)c4ccccc4F |
| Molecular Weight (g/mol) | 409.43 |
| Percent Purity | >95 |