Learn More
[3-[[[2-(Diaminomethyleneamino)-4-thiazolyl]methyl]thio]propionyl]sulfamide Hydrochloride, TRC
Product Code. 30137515
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137515 100 mg 100mg
17977704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137515 Supplier Toronto Research Chemicals Supplier No. TRCD416369100MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 76824-17-4
Molecular Formula C8 H14 N6 O3 S3 . Cl H
Molecular Weight (g/mol) 374.89
Synonym Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)-, hydrochloride (1:1), Propanamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)-, monohydrochloride (9CI), 3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]-N-sulfamoyl-propanamide Hydrochloride, Famotidine Imp. C (EP) as Hydrochloride
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N-sulfamoylpropanamide;hydrochloride
SMILES Cl.NC(=Nc1nc(CSCCC(=O)NS(=O)(=O)N)cs1)N
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C8H14N6O3S3.ClH/c9-7(10)13-8-12-5(4-19-8)3-18-2-1-6(15)14-20(11,16)17;/h4H,1-3H2,(H,14,15)(H2,11,16,17)(H4,9,10,12,13);1H
Formula Weight 374.0056
Percent Purity >95%
Chemical Name or Material 3-[([2-[(Diaminomethylene)amino]thiazol-4-yl]methyl)sulfanyl]-N-sulfamoyl-propanamide Hydrochloride
Grade HPLC