Learn More
3-((2-(Benzyloxy)phenyl)amino)cyclohex-2-enone, TRC
Specifications
Specifications
| Chemical Name or Material | 3-((2-(Benzyloxy)phenyl)amino)cyclohex-2-enone |
| CAS | 2817148-13-1 |
| Molecular Formula | C19 H19 N O2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C19H19NO2/c21-17-10-6-9-16(13-17)20-18-11-4-5-12-19(18)22-14-15-7-2-1-3-8-15/h1-5,7-8,11-13,20H,6,9-10,14H2 |
| SMILES | O=C1CCCC(=C1)Nc2ccccc2OCc3ccccc3 |
| IUPAC Name | 3-(2-phenylmethoxyanilino)cyclohex-2-en-1-one |
| Molecular Weight (g/mol) | 293.36 |
| Formula Weight | 293.142 |